missing translation for 'onlineSavingsMsg'
Learn More
1-Aminopyrene, 97%, Thermo Scientific Chemicals Product Code.: 15109153

1-Aminopyrene, 97%, Thermo Scientific Chemicals

Product Code. 15109153
5 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 15109153

Brand: Thermo Scientific Acros 104530050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1606-67-3
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.27
MDL Number MFCD00004140
InChI Key YZVWKHVRBDQPMQ-UHFFFAOYSA-N
Synonym 1-aminopyrene, 1-pyrenamine, 3-aminopyrene, pyrene, amino, pyren-1-ylamine, aminopyrene, pyrenamine, alpha-aminopyrene, unii-luw9eo1681, ccris 760Show MoreShow Less
PubChem CID 15352
IUPAC Name pyren-1-amine
SMILES NC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 12,1341
Formula Weight 217.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Aminopyrene, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454829 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number104530050,104530010,104532500
Lot NumberA0451715 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number104530050,104530010,104532500
Lot NumberA0400159 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number104530050,104530010,104532500
Lot NumberA0396109 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number104530050,104530010,104532500
Lot NumberA0410591 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number104530050,104530010,104532500
compliance-icons
Product Identifier
  • 1-Aminopyrene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Aminopyrene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.