Learn More
Labcon™ PerformR™ 15 mL Centrifuge Tubes
These centrifuge tubes are complete with graduations and a writing surface.
24.50 € - 329.00 €
Product Code | Brand | Closure Type | Material | Sterility | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Material | Sterility | Quantity | Price | Quantity & Availability | ||
16674042
|
Labcon™
3146-335-016-9 |
Flat Screw Cap | Polypropylene Resin | Sterile | 25 Tubes/Reusable Polypropylene Rack, 2 Racks/Pk., 6 Pk./Cs. |
111.00 €
Pack of 300 |
In Stock
Log in to see stock. |
||
16654032
|
Labcon™
3136-345-006-9 |
Flat Screw Cap | Polypropylene Resin | Sterile | 50 Tubes/Bag, 10 Bags/Cs. |
131.00 €
Pack of 500 |
In Stock
Log in to see stock. |
||
16684042
|
Labcon™
3146-870-006-9 |
Flat Screw Cap | Polypropylene Resin | Non-sterile | 500 Tubes/Bag, 500 Caps in Separate Bag, 500 Tubes and Caps/Cs. |
142.00 €
Pack of 500 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||
16674032
|
Labcon™
3140-335-026-9 |
Flat Screw Cap | Polystyrene Resin | Sterile | 25 Tubes/Low Waste Fiber Rack, 2 Racks/Pk., 10 Pk./Cs. |
329.00 €
Pack of 500 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||
16674022
|
Labcon™
3130-345-006-9 |
Flat Screw Cap | Polystyrene Resin | Sterile | 50 Tubes/Bag, 10 Bags/Cs. |
256.00 €
Pack of 500 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||
17275694
|
Labcon™
3128-355-026-9 |
Flat Screw Cap | Polystyrene | Sterile | 125 Tubes/Bag, 4 Bags /Cs. |
64.80 €
Pack of 125 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
17597896
|
Labcon™
3111-800-020-9 |
Flat Screw Cap | Polystyrene | Non-sterile | 1000 Tubes/Pk. |
164.00 €
Pack of 1000 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
17265694
|
Labcon™
3128-350-026-9 |
Flat Screw Cap | Polystyrene | Non-sterile | 125 Tubes/Bag, 4 Bags /Cs. |
57.70 €
Pack of 125 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||
16664032
|
Labcon™
3137-345-006-9 |
Plug Style Screw Cap | Polypropylene Resin | Sterile | 50 Tubes/Bag, 10 Bags/Cs. |
123.00 €
Pack of 500 |
In Stock
Log in to see stock. |
||
16694042
|
Labcon™
3147-870-006-9 |
Plug Style Screw Cap | Polypropylene Resin | Non-sterile | 500 Tubes/Bag, 500 Caps in Separate Bag, 500 Tubes and Caps/Cs. |
149.00 €
Pack of 500 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||
17517906
|
Labcon™
3147-335-016-9 |
Plug Style Screw Cap | Polystyrene Resin | Sterile | 25 Tubes/Rack, 2 Racks/Pk. |
24.50 €
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
Description
- Conical centrifuge tube
- ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
- Made in USA
Specifications
15 mL | |
Yes |