missing translation for 'onlineSavingsMsg'
Learn More
2-Cyclohexylethylamine, 97%, Thermo Scientific Chemicals Product Code.: 11307559

2-Cyclohexylethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11307559
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11307559

Brand: Thermo Scientific Alfa Aesar H27987.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Protect from direct light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4442-85-7
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00058668
InChI Key HFACYWDPMNWMIW-UHFFFAOYSA-N
Synonym 2-cyclohexylethylamine, 2-cyclohexyl-ethylamine, cyclohexaneethanamine, cyclohexaneethylamine, 2-cyclohexylethan-1-amine, cyclohexaneethanamine 9ci, 2-aminoethylcyclohexane, ethylamine, 2-cyclohexyl, chembl19428, 2-cyclohexylethyl amine hydrochlorideShow MoreShow Less
PubChem CID 20509
IUPAC Name 2-cyclohexylethanamine
SMILES NCCC1CCCCC1

Specifications

Quantity 5 g
UN Number UN2735
Sensitivity Air sensitive
Formula Weight 127.23
Percent Purity 97%
Chemical Name or Material 2-Cyclohexylethylamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10207381 Certificate TypeCertificate of Analysis Date20/01/2023 Catalog NumberH27987.03,H27987.06
Lot Number10243143 Certificate TypeCertificate of Analysis Date05/09/2022 Catalog NumberH27987.03,H27987.06
Lot Number10194823 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27987.03,H27987.06
Lot Number10156631 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27987.03,H27987.06
Lot Number10180638 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27987.03,H27987.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Cyclohexylethylamine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.