missing translation for 'onlineSavingsMsg'
Learn More
4-(2-Chlorophenoxy)benzenesulfonyl chloride, 96%, Thermo Scientific™ Product Code.: 15415067

4-(2-Chlorophenoxy)benzenesulfonyl chloride, 96%, Thermo Scientific™

Product Code. 15415067
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15415067

Brand: Thermo Scientific Alfa Aesar H32346.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610277-84-4
Molecular Formula C12H8Cl2O3S
Molecular Weight (g/mol) 303.15
MDL Number MFCD01631899
InChI Key ILZBGLOVCXTRDS-UHFFFAOYSA-N
Synonym 4-2-chlorophenoxy benzenesulfonyl chloride, 4-2-chloro-phenoxy-benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-chlorophenoxy, 4-2-chlorophenoxy benzene-1-sulfonyl chloride, chloro 4-2-chlorophenoxy phenyl sulfone, 4-2-chlorophenoxy phenylsulfonyl chlorideShow MoreShow Less
PubChem CID 2794710
SMILES ClC1=CC=CC=C1OC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 4-(2-chlorophenoxy)benzenesulfonyl chloride
Formula Weight 303.17
Percent Purity 96%
Chemical Name or Material 4-(2-Chlorophenoxy)benzenesulfonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10161142 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(2-Chlorophenoxy)benzenesulfonyl chloride, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.