missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-(trifluoromethoxy)benzoic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 221.13 |
Percent Purity | 95% |
Chemical Name or Material | 4-Amino-3-(trifluoromethoxy)benzoic acid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Amino-3-(trifluoromethoxy)benzoic acid, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction