missing translation for 'onlineSavingsMsg'
Learn More
4-Chlorobenzoyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 11384749

4-Chlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11384749
25 g
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Product Code. 11384749

Brand: Thermo Scientific Alfa Aesar A16325.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.

Solubility
Reacts with water. Reacts with alcohol.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alcohol, bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 122-01-0
Melting Point 12°C to 14°C
Density 1.377
Boiling Point 221°C to 222°C
Flash Point 105°C
Odor Acrid
Molecular Formula C7H4Cl2O
Refractive Index 1.578
MDL Number MFCD00000686
Quantity 25 g
UN Number UN3265
Beilstein 471606
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride
Solubility Information Reacts with water. Reacts with alcohol.
InChI Key RKIDDEGICSMIJA-UHFFFAOYSA-N
SMILES ClC(=O)C1=CC=C(Cl)C=C1
IUPAC Name 4-chlorobenzoyl chloride
Molecular Weight (g/mol) 175.01
PubChem CID 8501
ChEBI CHEBI:60716
Formula Weight 175.01
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzoyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238523 Certificate TypeCertificate of Analysis Date17/11/2023 Catalog NumberA16325.22,A16325.14,A16325.36
Lot Number10236040 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog NumberA16325.22,A16325.14,A16325.36
Lot Number10150432 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16325.22,A16325.14,A16325.36
Lot Number10103525 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16325.22,A16325.14,A16325.36
Lot Number10210480 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16325.22,A16325.14,A16325.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chlorobenzoyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.