missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[6-(Diethylamino)-3-pyridinyl]methanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 180.25 |
Percent Purity | 97% |
Chemical Name or Material | [6-(Diethylamino)-3-pyridinyl]methanol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
[6-(Diethylamino)-3-pyridinyl]methanol, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction