Learn More
8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech., Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 75.0 |
CAS Max % | 100.0 |
Color | Brown to Yellow |
Loss on Drying | 4 % to 6 % (1 g, 150°C, 4 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 75% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Synonym | h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt |
Solubility Information | Solubility in water: slightly soluble. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468848 | Certificate of Analysis | 31/01/2025 | 400841000,400845000,400840050 |
A0463013 | Certificate of Analysis | 20/05/2024 | 400841000,400845000,400840050 |
A0459546 | Certificate of Analysis | 31/01/2024 | 400841000,400845000,400840050 |
A0456283 | Certificate of Analysis | 29/09/2023 | 400841000,400845000,400840050 |
A0453443 | Certificate of Analysis | 15/09/2023 | 400841000,400845000,400840050 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.