missing translation for 'onlineSavingsMsg'
Learn More
9-Methylanthracene, 99%, Thermo Scientific Chemicals Product Code.: 11497657

9-Methylanthracene, 99%, Thermo Scientific Chemicals

Product Code. 11497657
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11497657

Brand: Thermo Scientific Alfa Aesar A15699.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.

Solubility
Soluble in water (0,0003 mg/L at 25°C) and toluene. Slightly soluble in benzene.

Notes
Mechanical exhaust required. When not in use, tightly seal the container and store in a dry, cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 779-02-2
Molecular Formula C15H12
Molecular Weight (g/mol) 192.26
MDL Number MFCD00001261
InChI Key CPGPAVAKSZHMBP-UHFFFAOYSA-N
Synonym anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standardShow MoreShow Less
PubChem CID 13068
IUPAC Name 9-methylanthracene
SMILES CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 77°C to 79°C
Density 1.066
Boiling Point 196°C to 197°C (16 mmHg)
Quantity 5 g
UN Number UN3077
Beilstein 1907463
Solubility Information Soluble in water (0,0003mg/L at 25°C) and toluene. Slightly soluble in benzene.
Formula Weight 192.26
Percent Purity 99%
Chemical Name or Material 9-Methylanthracene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251836 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15699.06,A15699.03,A15699.14
Lot Number10241917 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog NumberA15699.06,A15699.03,A15699.14
Lot Number10205308 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15699.06,A15699.03,A15699.14
Lot Number10215630 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15699.06,A15699.03,A15699.14
Lot Number9007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15699.06,A15699.03,A15699.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9-Methylanthracene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.