Learn More
3M™ Multi-Format Chemical Sorbent
Effectively controls aggressive liquid spills including acids and alkalis. 3M™ Multi-format Chemical Sorbent combines four formats in one product; mini-boom, pillow, sheet and roll.
Brand: 3M™ 7000002024
This item is not returnable.
View return policy
Description
Effective clean up and control of aggressive liquid spills including acids and alkalis. The dispenser combines four formats in one product; mini-boom, pillow, sheet and roll.- Sheets are perforated at 40cm intervals
- Can be used with many aggressive chemicals
- Immediate response to chemical spillages for fast and effective containment and clean-up
- Sorbent materials can be disposed of easily.
Specifications
Yellow | |
Multi-format Chemical Sorbents | |
1 Ea. | |
120 mm |
119 L | |
Cleaning and containing chemical-based spills such as acids/alkalis/alcohols, chemicals, flammable liquids, and other solvents. | |
15.2 m | |
8.2 kg |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Product Title
3M™ Multi-Format Chemical Sorbent > Multi-Format Chemical Sorbents
Spot an opportunity for improvement?Share a Content Correction