Learn More
Cole-Parmer™ Aluminum Tensioning Tools
Adjust tension from light to very tight using a knob at the base of this tool. Cole-Parmer™ Aluminum Tensioning Tool cuts ties automatically when preset tension is reached. Tension knob features lock to ensure repeatable tension, tie after tie.
Brand:  Cole-Parmer™ 06830-00
Product Code. 11762753
  • 471.00 € / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Adjust tension from light to very tight using a knob at the base of this tool. Cole-Parmer™ Aluminum Tensioning Tool cuts ties automatically when preset tension is reached. Tension knob features lock to ensure repeatable tension, tie after tie.
Shock-resistant tool absorbs all impact. Ideal for 18 to 175 lb. cable or zip ties. No twisting necessary.
Specifications
Specifications
Tensioning Tools
Aluminum
1 Ea.
Cable, Zip Ties, Aluminum Gun, Stiffer
The Aluminum Gun, Stiffer Is Recommended For Jobs in Large Series
−40° to +85°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Aluminum Tensioning Tools > Aluminum