Learn More
Thermo Scientific™ ART™ SolventSafe Pipette Tips
GREENER_CHOICE
Perform applications accurately and consistently with these Solventsafe pipette tips, which seal when exposed to potential contaminants.
Brand:  Thermo Scientific™ 5479E
92.20 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 10252431
  • 116.40 € / Pack of 400
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ SolventSafe Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ART SolventSafe 1000E, Filtered, Sterile, Rack
Filtered
ART SolventSafe 1000E
8.83cm
Micro Point
ART
1000 μL
Sterile
3.47 in.
Rack
Universal Fit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ ART™ SolventSafe Pipette Tips > ART Barrier Pipette tips SolventSafe 1000E; 1000μL MicroPoint, sterile tip