missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 96-well PCR Microplates

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.
45.07 € - 393.58 €
Specifications
Barcode | No Barcode |
---|---|
Certifications/Compliance | RNase-, DNase-free |
Array | 8 x 12 |
Material | Polypropylene |
Volume (Metric) Well | 200 μL |
Product Code | Brand | Color | Skirt Style | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Skirt Style | Format | Price | Quantity & Availability | |||||
11396124
![]() |
Axygen™
PCR-96-AB-C |
Clear | Elevated | Standard | N/A | ||||||
12799438
![]() |
Axygen™
PCR-96-AB-C |
Clear | Elevated | Standard |
314.09 €
Pack of 50 |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
11391804
|
Axygen™
PCR-96-C |
Clear | Unskirted | Standard | N/A | ||||||
12112580
|
Axygen™
PCR-96-C |
Clear | Unskirted | Standard |
263.09 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
11330185
|
Axygen™
PCR-96-FLT-C |
Clear | Unskirted | Flat Top | N/A | ||||||
12335193
|
Axygen™
PCR-96-FLT-C |
Clear | Unskirted | Flat Top |
393.58 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
12719458
|
Axygen™
PCR-96-FS-C |
Clear | Full | Standard |
269.51 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
11380185
|
Axygen™
PCR-96-FS-C |
Clear | Full | Standard | N/A | ||||||
12799448
|
Axygen™
PCR-96-FS-B |
Blue | Full | Standard |
341.51 €
Pack of 50 |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
12779458
|
Axygen™
PCR-96-FS-Y |
Yellow | Full | Standard | N/A | ||||||
12739458
|
Axygen™
PCR-96-FS-O |
Orange | Full | Standard | N/A | ||||||
12779448
|
Axygen™
PCR-96-FLT-W |
White | Unskirted | Flat Top | N/A | ||||||
12749458
|
Axygen™
PCR-96-FS-R |
Red | Full | Standard | N/A | ||||||
12709458
|
Axygen™
PCR-96-FS-BK |
Black | Full | Standard | N/A | ||||||
12789448
|
Axygen™
PCR-96-FS-A |
Assorted | Full | Standard | N/A | ||||||
12799478
![]() |
Axygen™
PCR-96-MB-C |
Clear | Unskirted | Elevated Wells | N/A | ||||||
10332390
|
Axygen™
PCR-96-HS-C |
Clear | Half | Standard |
294.68 €
Pack of 50 |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
12537797
|
Axygen™
PCR-96M2-HS-C |
Clear | Half | Single Notch Corner | N/A | ||||||
13164824
|
Axygen™
PCR-96M2-HS-C |
Clear | Half | Single Notch Corner |
314.09 €
Pack of 50 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
12769458
|
Axygen™
PCR-96-FS-W |
White | Full | Standard | N/A | ||||||
Description
Axygen™ 96 well PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.- RNase-/DNase-free
- Nonpyrogenic
- Nonsterile
- Manufactured with 99.9% pure virgin polypropylene
- Single Notch Coner
- Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
- Well Volume: 200 μL
Specifications
No Barcode | |
8 x 12 | |
200 μL | |
Non-sterile |
RNase-, DNase-free | |
Polypropylene | |
96 | |
PCR thermal transfer, RT-PCR applications |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ 96-well PCR Microplates