Learn More
Axygen™ 96-well PCR Microplates
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.
Brand:  Axygen™ PCR-96-C
 View more versions of this product
Product Code. 12112580
  • 263.09 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen™ 96 well elevated skirt PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96-well Semi-Skirted PCR Plate
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    No Barcode
    Standard
    8 x 12
    Statagene MX4000
    Polypropylene
    96
    Round
    50/Cs.
    RNase-, DNase-free
    Without Lid
    Clear
    Unskirted
    200 μL
    Non-sterile
    PCR thermal transfer, RT-PCR applications
    10/Pk.
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Axygen™ 96-well PCR Microplates > Clear; No Skirt; 96-well; 200 μL