Learn More
BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems
Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies provide catalyst-free convenience.
Brand:  BD 260683
 View more versions of this product
Product Code. 11777194
  • 185.00 € / Pack of 20
Explore more special offers
Description
Description
  • One-step resealable pouches
  • The pouches are specially designed to maximize the preservation of the desired environment throughout incubation
  • This heat-free technology minimizes condensate, allowing excellent bacterial colony visualization though the pouch
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EZ Anaerobe Pouch System
Resealable Pouches
BD GasPak EZ Small Incubation Container
GasPak
Ziplock
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Resealable pouches with indicator
20 Sachets
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems > Anaerobe Pouch System