missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 96-well Deep-well Plates
These deep-well plates are stackable for easy storage.
Brand: BRAND™ 701355
368.64 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- DNA-, DNase- and RNase-free
- Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80°C
- Deep-well plates are manufactured according to ANSI/SLAS 1 and 4 in.
- Choose PP plates for autoclavability and chemical resistance.
- Choose PS plates for transparency and surface properties.
- Deep Well Plates are ideal for sample storage, HTS assays requiring a motherplate, cell and tissue culture, immunological assays, and other applications.

Specifications
Standard | |
Autoclavable | |
Polypropylene | |
384 | |
Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination | |
U-bottom |
16 x 24 | |
Deep-well Plate | |
0.3 mL | |
Round Bottom (U-well) | |
48 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
052556 | Certificate of Analysis | 14/10/2024 | 701355 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ 96-well Deep-well Plates > Well Volume: 0.3 mL, Material: Polypropylene, Bottom: Round
Spot an opportunity for improvement?Share a Content Correction