missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ CENTRY™ 101 Plate Centrifuge

Proprietary swing-out rotor design prevents microplate contents from spilling, increasing the reliability of your microplate reactions. The CENTRY™ 101 Plate Centrifuge accepts popular PCR plates and most microplates.

Brand:  Gilson™ 36117410

 View more versions of this product

Product Code. 16358641

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Decreased spill risk:

  • The CENTRY™ 101 Plate Centrifuge offers a patented swing-out rotor design that keeps the microplate contents from spilling out while concentrating all reaction components in the well bottom, improving the accuracy of your reactions, such as sensitive PCR reactions.
  • The rotor carriers rest at a 75° angle, allowing you to insert plates without sealing tape or caps.
  • At speed, the microplates run vertically.
  • The two-plate capacity CENTRY™ 101 Plate Centrifuge accommodates most PCR and microplates.

Small, but fast:

  • Samples are spun down in less than 20 seconds at 2550 rpm (600 x g).
  • With a small footprint (23 x 26 cm, 9.2 x 10.2 in.), the CENTRY™ 101 Plate Centrifuge easily fits into hoods and on crowded benchtops.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2,550 rpm
UK-plug
220/230 V
23 x 26 x 19.7 cm
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilson™ CENTRY™ 101 Plate Centrifuge > CENTRY™ 101 Plate Centrifuge; Type: UK Plug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.