Learn More
Certified Reference Material Distillation Standard, Gasoline, UKAS ISO 17034 Certified 250mL (nominal value: 26.4 °C to 174.9 °C), Paragon Scientific™
Distillation Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Distillation according to ASTM D86. Alternative test methods are IP 123 and ISO 3405.
Brand: Paragon Scientific CRMU-DIGA
Description
- Fully certified in accordance with ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
- Low levels of uncertainty achieves maximum accuracy of data
- All data generated exclusively by UKAS ISO 17025 accredited laboratories
- Fully traceable to international standards
- Supplied in tamper evident UN compliant security pack
- 12 month shelf life
- Available from stock for immediate dispatch
- Manufactured in the United Kingdom
Distillation Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Distillation according to ASTM D86. Alternative test methods are IP 123 and ISO 3405.
Specifications
250 mL | |
UN 1203 |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- 250ML CRMU-DIGA Distillation Standard, Gasoline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H360D-May damage the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P370+P378-In case of fire: Use media other than water to extinguish.
Supplemental information
- MIXTURE LIST-Contains : gasoline
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Nominal values quoted. Certified Values will vary from batch to batch.