missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chromogenic Coliform Agar (Dehydrated)

Detect, enumerate and differentiate coliforms and E.coli in water samples using Chromogenic Coliform Agar in accordance with ISO 9308-1.

Brand:  Thermo Scientific™ CM1205B

878.75 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.


Product Code. 15744047

  • 925.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Enumerate and differentiate coliforms and Escherichia coli in water samples with Thermo Scientific™ Oxoid™ Chromogenic Coliform Agar in accordance with ISO 9308-1 guidelines for water quality analysis. Target organisms are easy to differentiate, E.coli forming dark blue/violet colonies and coliforms pink to red colonies. Achieve complete detection within 24 hours, removing the need for subculturing.

Chromogenic Coliform Agar is a fast, accurate, and efficient way to detect coliforms and E.coli during microbiological quality testing of drinking water, disinfected pool water, and treatment plant samples. The guideline, ISO 9308-1:2014 Enumeration of Escherichia coli and coliform bacteria –Part 1: Membrane filtration method for waters with low bacterial background flora, specifies a method based on membrane filtration, subsequent culture on a Chromogenic Coliform Agar medium, and calculation of the number of target organisms in the sample.

Chromogenic Coliform Agar utilizes specific enzymes associated with general coliforms and E.coli for differentiation.

  • The chromogen Salmon GAL, when cleaved by D-galactosidase, produces a pink to red coloration; this coloration and enzyme are associated with coliforms.
  • E.coli contains D-galactosidase, which cleaves Salmon GAL; however, it also contains D-glucuronidase, which cleaves the chromogen X-glucuronide. The combination of these chromogens produces a dark blue to violet coloration associated with E.coli.
  • Tergitol-7 within the medium inhibits Gram positive bacteria and Gram negative bacteria with exception of coliforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10°C to 30°C
Powder
500 g
Chromogenic Coliform Agar (Dehydrated)
Agar
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6254810 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog NumberCM1205B
Lot Number6183394 Certificate TypeCertificate of Analysis Date16/11/2024 Catalog NumberCM1205B
Lot Number6137481 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberCM1205B
Lot Number6117213 Certificate TypeCertificate of Analysis Date25/05/2024 Catalog NumberCM1205B
Lot Number6102791 Certificate TypeCertificate of Analysis Date10/05/2024 Catalog NumberCM1205B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Chromogenic Coliform Agar (Dehydrated) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.