Learn More
Thermo Scientific™ Chromogenic Coliform Agar (ISO)
Detect, enumerate, and differentiate coliforms and E.coli in water samples in accordance with ISO 9308-1 with convenient easy-to-read prepared media.
Brand: Thermo Scientific™ PO5318A
Description
Enumerate and differentiate coliforms and Escherichia coli in water samples with Thermo Scientific™ Oxoid™ Chromogenic Coliform Agar (ISO) prepared media plates, in accordance with ISO 9308-1 guidelines for water quality analysis. Target organisms are easy to differentiate, E.coli forming dark blue/violet colonies and coliforms pink to red colonies. Complete detection within 24 hours, removing the need for subculturing.
Chromogenic Coliform Agar (ISO) plates are a fast, accurate, and efficient way to detect coliforms and E.coli during microbiological quality testing of drinking water, disinfected pool water, and treatment plant samples.- Conforms to ISO 9308-1 guidelines
- Easily and clearly differentiates E. coli and coliforms
- Ready-to-use convenience of prepared media
Chromogenic Coliform Agar (ISO) utilizes specific enzymes associated with general coliforms and E.coli for differentiation.
- The chromogen Salmon GAL, when cleaved by D-galactosidase, produces a pink to red coloration; this coloration and enzyme are associated with coliforms.
- E.coli contains D-galactosidase which cleaves Salmon GAL; however, it also contains D-glucuronidase, which cleaves the chromogen X-glucuronide. The combination of these chromogens produces a dark blue to violet coloration associated with E.coli.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Light ivory, transparent | |
Chromogenic Coliform Agar (ISO) | |
Germany | |
17 mL | |
Flow Wrapped - Single Layer |
2° to 12°C, in the dark | |
90mm Plate | |
Agar | |
17mL | |
10/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6252092 | Certificate of Analysis | 01/04/2025 | PO5318A |
6249190 | Certificate of Analysis | 21/03/2025 | PO5318A |
6249191 | Certificate of Analysis | 21/03/2025 | PO5318A |
6246003 | Certificate of Analysis | 17/03/2025 | PO5318A |
6246004 | Certificate of Analysis | 17/03/2025 | PO5318A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.