Learn More
Eppendorf™ Concentrator Plus Rotors
Designed with the customer in mind to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench
Brand:  Eppendorf™ 5490043006
590.73 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 10609503
  • 563.24 € / Each
Explore more special offers
Description
Description
  • Advanced heating technology provides best treatment for your sample which assures quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. Our new coated lid provides superior chemical resistance against aggressive acids and organic solvents (e.g. TFA, DMSO).
  • The Concentrator plus is available with three configurations to meet the customers' lab specific needs:
  • Basic Concentrator for connecting to an existing vacuum pump
  • Complete system with an integrated solvent-resistant, maintenance and oil-free PTFE pump, and condensation trap
  • Complete system with vacuum port adds extra capability. It allows for using the integrated vacuum pump separately – without having to disconnect it from the system
  • Choice of 3 operation modes (vacufuge, centrifuge, and desiccator) for a variety of application needs
  • Choice of three application modes (aqueous, alcohol or high vapor pressure) to correspond with sample solvent—reduction of processing time up to 20%
  • Choice of 4 heating levels (ambient, 30C, 45°C, 60°C) allows safe and efficient concentration of various samples
  • Brake function can optionally be turned off for use with delicate samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor F-45-16-20
16 x 6.5/10 mL
20 mm
Vertrieb Rotor
55 mm
Concentrator Plus Centrifuge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Concentrator Plus Rotors > Capacity: 16 x 6.5/10mL