Learn More
WLD Tec™ Safety Adapter for CV360 Gas Cartridge
Suitable and safe gas cartridge adapter for Gasprofi 1 SCS laboratory gas burners. WLD Tec™ Safety Adapter for CV360 Gas Cartridge includes and integrated pressure regulator.
Brand: WLD Tec™ 8.000.700
This item is not returnable.
View return policy
Description
- Safety adapter
- Includes integrated pressure regulator
- Accepts gas cartridges, CV 360
Specifications
Gas Adapter | |
Gasprofi 1 SCS Laboratory Gas Burners | |
CV 360 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
WLD Tec™ Safety Adapter for CV360 Gas Cartridge > Gas type: CV 360
Spot an opportunity for improvement?Share a Content Correction