Learn More
Invitrogen™ Cytochalasin D
Cytochalasin D is a cell permeable fungal toxin that binds to the barbed end of actin filaments inhibiting both the association and dissociation of subunits.
Brand: Invitrogen™ PHZ1063
Description
Cytochalasin D is a cell permeable fungal toxin that binds to the barbed end of actin filaments inhibiting both the association and dissociation of subunits. This compound causes the disruption of actin filaments and inhibition of actin polymerization.
Format/Formulation: Lyophilized

Specifications
Store in freezer (-5°C to -30°C). | |
Dry Ice | |
Non-sterile | |
Cytochalasin D | |
507.6 | |
Solid |
60 | |
Actin | |
1 mg | |
Chemical | |
Cytoskeletal Inhibitor |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- CYTOCHALASIN D INHIBITOR
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P361-Take off immediately all contaminated clothing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
77344123 | Certificate of Analysis | 29/01/2025 | PHZ1063 |
77260032 | Certificate of Analysis | 29/01/2025 | PHZ1063 |
77465605 | Certificate of Analysis | 29/01/2025 | PHZ1063 |
77530366 | Certificate of Analysis | 29/01/2025 | PHZ1063 |
77683108 | Certificate of Analysis | 29/01/2025 | PHZ1063 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.