missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D(+)-Proline, ≥99% Product Code.: 10470501

Thermo Scientific Chemicals D(+)-Proline, ≥99%

Product Code. 10470501
5 g, Glass Bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10470501

Brand: Thermo Scientific Chemicals 228660050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00064317
InChI Key ONIBWKKTOPOVIA-SCSAIBSYSA-N
Synonym d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-prolineShow MoreShow Less
PubChem CID 8988
ChEBI CHEBI:16313
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 1 % max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Glass Bottle
Beilstein 22, 2
Specific Rotation +83.0° to +87.0° (20°C, 589nm) (c=4, water)
Fieser 07,307; 09,393
Sulfated Ash 0.1 % max.
Specific Rotation Condition +84.75 (20.00°C c=4,H2O)
Quantity 5 g
Formula Weight 115.13
Percent Purity 99.0 to 101.0% (Total base)
Physical Form Powder
Chemical Name or Material D(+)-Proline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464636 Certificate TypeCertificate of Analysis Date29/07/2024 Catalog Number228660050,228660010,228660250
Lot NumberA0424563 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number228660050,228660010,228660250
Lot NumberA0400967 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number228660050,228660010,228660250
Lot NumberA0396602 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number228660050,228660010,228660250
Lot NumberA0399758 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number228660050,228660010,228660250
Product Identifier
  • D(+)-Proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D(+)-Proline, ≥99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.