missing translation for 'onlineSavingsMsg'
Learn More
D-Proline, 99%, Thermo Scientific Chemicals Product Code.: 11493444

D-Proline, 99%, Thermo Scientific Chemicals

Product Code. 11493444
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11493444

Brand: Thermo Scientific Alfa Aesar A10682.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.

Solubility
Soluble in water (1620 g/L at 20°C), and acetone (slightly). Insoluble in ethanol.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00064317
InChI Key ONIBWKKTOPOVIA-SCSAIBSYSA-N
Synonym d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
PubChem CID 8988
ChEBI CHEBI:16313
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Melting Point 213°C to 215°C (decomposition)
Beilstein 80811
Solubility Information Soluble in water (1620g/L at 20°C),and acetone (slightly). Insoluble in ethanol.
Density 1.36
Quantity 5 g
Formula Weight 115.13
Optical Rotation +84° (c=1 in water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material D-Proline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245314 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog NumberA10682.06,A10682.03,A10682.14
Lot Number10241625 Certificate TypeCertificate of Analysis Date18/07/2022 Catalog NumberA10682.06,A10682.03,A10682.14
Lot Number10238029 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10682.06,A10682.03,A10682.14
Lot Number10099566 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10682.06,A10682.03,A10682.14
Lot Number10113828 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10682.06,A10682.03,A10682.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Proline, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.