missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Sucrose (Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
Description
D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation.

Specifications
Specifications
Alpha Vector | Sucrose |
Melting Point | 190°C |
Color | White |
pH | 6.5 to 7.5 |
Ignition Residue | 0.01% max. |
Loss on Drying | 0.03% max. |
Assay Percent Range | ≥99.9% |
Packaging | Poly Pail |
Quantity | 10 kg |
Merck Index | 15, 9012 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
D-Sucrose (Molecular Biology), Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction