Learn More
Fisherbrand™ Deionization Cartridges
Purify water with single use cartridges. Fisherbrand™ Deionization Cartridges change color when saturation is detected.
Brand:  Fisherbrand™ R3
88.45 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 11954159
  • 94.45 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Detects the saturation by color change
  • Direct connection to the water network
  • A wide choice of cartridges to meet all needs with reduced cost
  • No electrical connection
  • A transparent building to monitor the saturation of the resin without auxiliary measurement
  • The disposable cartridges contain only new resins. The performance level is constant and guaranteed
  • At saturation of the resin cartridges are simply discarded
  • The small dimensions allow easy installation
  • Applications: dishwashing, cooling circuit filling, battery, battery and radiator antifreeze preparation, photo development
Specifications
Specifications
Blue cartridge turns ocher brown
20 L/hr.
500 mm
For R3 series
1 Each
10°C to 35°C
Depends on the quality of the inlet water
Ion Exchanger
For water purification, sensitive applications
3 bar
35°C
Cartridge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Deionization Cartridges > For R3