missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DMEM/F-12

DMEM/F-12
Brand: Gibco™ 11320074
15.39 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM / F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM / F-12 product made in our Grand Island facility (11320-033). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
1 X | |
Dulbecco's Modified Eagle Medium | |
No HEPES | |
500 mL | |
7 to 7.4 | |
2 to 8°C |
Liquid | |
Glutamine, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate | |
For mammalian cell culture | |
ISO 13485 standards, FDA | |
CE-IVD | |
Dulbecco's Modified Eagle Medium, 500mL, Liquid Form, 1 X Concentration, 7 to 7.4 pH, 2 to 8°C Storage Temperature, 12 Months Shelf Life, Certified to ISO 13485 Standards, For Supporting Growth of Mammalian Cells |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 13
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 2,6-Dichlorobenzoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 2,6-Dichlorobenzoicacid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ DMEM/F-12 > 500mL
Spot an opportunity for improvement?Share a Content Correction