missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DMEM/F-12, GlutaMAX™ Supplement

DMEM/F-12, GlutaMAX™ Supplement
Brand: Gibco™ 31331028
25.37 EUR valid until 2025-12-31
Use promo code "24331" to get your promotional price.
This item is not returnable.
View return policy
Description
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.
cGMP manufacturing and quality system
DMEM/F-12 is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical DMEM/F-12 product made in our Scotland facility (31331-028). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard.

Specifications
1 X | |
DMEM/F-12 Medium | |
No HEPES | |
500 mL | |
6.8 to 7.2 | |
DMEM/F-12 Medium, 500mL, 1 X Concentration, Liquid Form, With GlutaMAX™, Phenol Red, Sodium Pyruvate, 6.8 to 7.2 pH, 12 Months Shelf Life, 290 to 330mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |
Liquid | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
Mammalian cell culture | |
ISO 13485 standards, FDA | |
2 to 8°C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ DMEM/F-12, GlutaMAX™ Supplement > 500mL
Spot an opportunity for improvement?Share a Content Correction