missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM/F-12, GlutaMAX™ supplement
GREENER_CHOICE

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.

Brand:  Gibco™ 10565018

34.62 EUR valid until 2025-12-31
Use promo code "24273" to get your promotional price.


 View more versions of this product

Product Code. 11559726

  • 38.27 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM / F-12 is modified as follows:

With: GlutaMAX, Phenol Red

Without: HEPES

The complete formulation is available.

Using DMEM/F-12

DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No HEPES
Gibco™, GlutaMAX™
Room Temperature
MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts
1 X
DMEM (Dulbecco's Modified Eagle Medium)/F-12
Sterile-filtered
High Glucose, GlutaMAX, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2581555 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10565018
Lot Number2646180 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10565018
Lot Number2581535 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10565018
Lot Number2537550 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10565018
Lot Number2581543 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10565018
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ DMEM/F-12, GlutaMAX™ supplement > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.