missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug

Made of borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug are used for transferring fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.

Brand:  Lenz Laborglasinstrumente™ 04477304

 View more versions of this product

Product Code. 11896144

  • 227.00 € / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Conical shape with NS sleeve and NS core
  • With PTFE plug
  • With threaded valve protection
  • Borosilicate glass
Specifications

Specifications

2000 mL
Borosilicate Glass
6.0 mm bore
PTFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug > Capacity (Metric): 2000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.