missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Untouched™ Mouse CD8 Cells Kit

Dynabeads™ Untouched™ Mouse CD8 Cells supplies both 4.5 μm superparamagnetic Dynabeads (with a polyclonal sheep anti-rat IgG antibody covalently bound to the bead surface) and a monoclonal Antibody Mix.

Brand:  Invitrogen™ 11417D

839.80 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10037862

  • 988.00 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dynabeads™ Untouched™ Mouse CD8 Cells supplies both 4.5 μm superparamagnetic Dynabeads (with a polyclonal sheep anti-rat IgG antibody covalently bound to the bead surface) and a monoclonal Antibody Mix, together designed for the depletion of CD8-negative leucocytes from mouse spleen or lymph node single-cell suspensions. Following the depletion procedure, the negatively isolated, untouched target mouse CD8+ cells are pure, viable, and ready to be used in any downstream application.

• Rapid isolation of untouched mouse CD8+ T cells—no columns required
• Pure and viable mouse CD8+ T cells are ready for any downstream application

Excellent yield, purity, and viability of recovered cells
The Antibody Mix contains a cocktail of rat IgGs that binds mouse CD4+T cells, B cells, NK cells, monocytes/macrophages, dendritic cells, erythrocytes, and granulocytes. The Antibody Mix is first added to the sample in a tube to bind the unwanted CD8-negative cells. The cells are then washed to remove excess antibodies. Mouse Depletion Dynabeads™ (coated with a rat IgG antibody) are added to bind the antibody-labeled, CD8-negative cells. After a short incubation, the bead-bound cells are then separated from unbound cells in 1–2 minutes using a magnet

The purified target mouse CD8+cells in the supernatant can be transferred to a new tube for use in any downstream application. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity, recovery and viability of the untouched CD8+ cells. The protocol can be scaled up according to your sample size.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 x 10 mL
Contains: 20 mL Mouse Depletion Dynabeads™ and 2 mL Antibody Mix with monoclonal antibodies towards CD4, CD11b, CD16/32, CD45R and Ter-119.

Store kit components at 2°C to 8°C.
Mouse
DYNAL™, Dynabeads™
Mouse
T Cells (CD8)
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2810556 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11417D
Lot Number2803107 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11417D
Lot Number2829965 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11417D
Lot Number2847711 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number11417D
Lot Number2880923 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number11417D
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Dynabeads™ Untouched™ Mouse CD8 Cells Kit > 2 x 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.