Learn More
Thermo Scientific™ EZ-Link™ HPDP-Biotin, No-Weigh™ Format
Brand: Thermo Scientific™ 21341
499.70 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ EZ-Link HPDP-Biotin is pyridyldithiol-activated, sulfhydryl-reactive biotinylation reagent that conjugates via a cleavable (reversible) disulfide bond to enable use in a variety of purification methods.
Highlights:
- Pyridyldithiol reacts with -SH groups to form a stable disulfide bond
- Absorbance of pyridine-2-thione leaving group at 343nm can be used to determine reaction progress
- Disulfide bond is cleavable using 50mM DTT (Part No. 20291) or 100mM β-Mercaptoethanol (Part No. 35600)
- Extended spacer arm prevents steric hindrance
- Must be dissolved in DMSO or DMF before use
- Biotinylation occurs between pH 6-9

Specifications
HPDP-Biotin | |
Store at 4°C. | |
Cell-Permeant | |
Pyridyldisulfide | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Thiol | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) | |
Long, Cleavable |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.