missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Agar, Powder / Flakes, Fisher BioReagents™

Used as a solidifying agent for culture media.

Brand:  Fisher BioReagents™ BP1423-500

88.87 EUR valid until 2025-12-31
Use promo code "24336" to get your promotional price.


 View more versions of this product

Product Code. 10572775

  • 93.55 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Used as a solidifying agent for culture media

  • Used as a solidfying agent to provide the solid medium to support the growth of microbes for cell culture applications
  • Functionally tested for promotion of growth of E. coli and S. aureus
  • Typical concentration is 1.5% agar (15g/L) in water
  • CAS: 9002-18-0
  • Molecular Weight: 336.337 g/mol
  • Chemical Formula: C14H24O9
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Powder to Flakes
Agar
Poly Bottle
Molecular genetics
Used for a solidifying agent for culture media
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher BioReagents™ Agar, Powder / Flakes, Fisher BioReagents™ > Poly bottle; Quantity: 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.