Learn More
Fisherbrand™ accumet™ Probe
Simple Plug and Stir technology for accumet XL and AB series meters
Brand:  Fisherbrand™ 01X478104
173.03 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 12860653
  • 220.00 € / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Simple Plug and Stir technology for accumet XL and AB series meters
Provides mixing without magnetic stir bars or stir plates required.
  • Saves valuable bench space
  • Quiet operation
  • Adjustable speed control
  • No additional power required
  • Multi-parameter XL benchtop models can accept up to two probes for simultaneous stirring
Order Info
Compatible with the following models: XL150, XL200, XL250, XL500, XL600, AB150, AB250, AB200. Replacement paddles available separately; see cat. no. 13620RP.
Services
One year
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Probe
Model XL150, 200, 250, 500, 600, AB150, 200 and 250 Multiparameter Benchtop Meter
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ accumet™ Probe > Stirring probe