missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ StemPro™ Accutase™ Cell Dissociation Reagent
StemPro Accutase Cell Dissociation Reagent is a cell detachment solution of proteolytic and collagenolytic enzymes.
Brand: Gibco™ A1110501
65.79 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
StemPro® Accutase® Cell Dissociation Reagent is a cell detachment solution of proteolytic and collagenolytic enzymes. Accutase® does not contain mammalian or bacterial derived products and is useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware, including Geltrex™, CELLStart™ and polymers. Accutase® performs exceptionally well in detaching primary and stem cells and maintaining high cell viabilty compared to animal origin enzymes such as trypsin. Some downstream applications following Accutase® treatment include analysis of cell surface markers, virus growth assay, cell proliferation, tumor cell migration assays, routine cell passage, production scale-up (bioreactor), and flow cytometry. Cell lines tested for Accutase® application include human embryonic stem cells, human mesenchymal stem cells, human neural stem cells and primary cells including macrophages, fibroblasts, keratinocytes, vascular endothelial cells, hepatocytes, and established cell lines such as adherent CHO and BHK cells and 293 cells. A complete list of published references is available on our website. Accutase® is a trademark of ICT, Inc.
Specifications
Store in freezer (-5 to -20°C) and protect from light. |
|
Serum-free | |
Frozen | |
Serum-Free | |
6 to 8 | |
Research Grade | |
Dry Ice |
Stem Cells | |
Adherent Cell Culture | |
Cell Culture Dissociation Reagent | |
Sodium Phosphate | |
Gibco™, StemPro™ | |
100 mL | |
Serum-Free |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 13
StemPro® Accutase® Cell Dissociation Reagent Brochure
File size: 676.6 kB
User Guide: StemPro Accutase Cell Dissociation Reagent
Version: B
File size: 153.5 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2442649 | Certificate of Analysis | 29/01/2025 | A1110501 |
2442657 | Certificate of Analysis | 29/01/2025 | A1110501 |
2442646 | Certificate of Analysis | 29/01/2025 | A1110501 |
2442655 | Certificate of Analysis | 29/01/2025 | A1110501 |
2442662 | Certificate of Analysis | 29/01/2025 | A1110501 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ StemPro™ Accutase™ Cell Dissociation Reagent > Quantity: 100 mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction