Learn More
Gilson™ PIPETMAN™ L Fixed Pipet
GREENER_CHOICE
The PIPETMAN L Fixed pipettes offer the same comfortable, lightweight design as the variable volume models, but the volume can never accidently be changed, making them an ideal choice for clinical diagnostics, quality control, and any routine testing applications.
Brand:  Gilson™ FA10019
 View more versions of this product
Product Code. 15134165
  • 226.00 € / Each
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Improved handle design for intensive users with lower weight and larger hook.
  • Graduated cover can be readjusted by users in the laboratory for dense and viscous fluids (once reset, the new volume cannot be accidentally changed).
  • Bottom part of the push button is colored in blue making it easy-to-identify as a fixed-volume model.
  • For a convenient access to the serviceable parts, the pipette is equipped with the Gilson clip ejector.
  • 2D identification code for easy traceability.
  • Tracking feature includes the product reference number, date of manufaturing, serial number, and nominal volume.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ PIPETMAN™ L Fixed Pipet > PIPETMAN™ L Fixed Pipet; Model: F5L; Volume: 5 μL; Color: Red