missing translation for 'onlineSavingsMsg'
Learn More
Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals Product Code.: 10823781

Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals

Product Code. 10823781
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10823781

Brand: Thermo Scientific Acros 364795000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH5N3·HCl
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochlorideShow MoreShow Less
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Absorbance (6M aq. soln.), (6 M aq. soln.) at 0nm, 0.01 max. at 280nm, 0.03 max. at 260nm
Melting Point 178.0°C to 188.0°C
Color Colorless to White
Infrared Spectrum conform to structure and standard
Assay Percent Range 99.5%
Packaging Plastic bottle
Linear Formula H2NC(=NH)NH2·HCl
Quantity 500 g
Beilstein 03,82
Merck Index 15,4597
Solubility Information Solubility in water: 2150g/L (20°C).
Formula Weight 95.53
Percent Purity 99.5%
Physical Form Crystals
Chemical Name or Material Guanidine hydrochloride, Without anticaking agent
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448536 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number364795000,364791000,364790100,364790025
Lot NumberA0462693 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number364795000,364791000,364790100,364790025
Lot NumberA0444616 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number364795000,364791000,364790100,364790025
Lot NumberA0448536 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number364795000,364791000,364790100,364790025
Lot NumberA0455153 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number364795000,364791000,364790100,364790025
compliance-icons
Product Identifier
  • Guanidine hydrochloride, without anticaking agent
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.