Learn More
Cytiva HiTrap™ MabSelect™ SuRe™ LX Columns
Polyclonal antibodies, monoclonal antibodies (mAb), and antibody fragments are usually purified by affinity chromatography. Resins containing an immobilized ligand (e.g., protein A, protein G, or protein L) are used to capture antibodies and antibody fragments.
Brand: Cytiva 29268402
Description
- Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
- High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
- Generic elution profile for different monoclonal antibodies enables platform approach to purification
- High-flow agarose matrix allows the processing of large feed volumes
- Reduces the risk of product contamination and carryover
- Improves performance and reduces overall costs

Specifications
Antibody Purification Column | |
4 mL/min. (Max) | |
85 μm | |
1 mL | |
1 to 30 mg/Column |
MabSelect SuRe LX | |
Protein A | |
Human IgG | |
Purification of Human IgG | |
5/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- HiTrap MabSelect SuRe LX
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.