Learn More
Thermo Scientific™ Immuno Breakable Modules Clear, C8 LockWell, MediSorp

Choose from four surfaces and a variety of wells and configurations. Immuno Breakable Modules offer an optimal choice of products for the specific assay.
Brand: Thermo Scientific™ 446470
502.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Choose from four surfaces and a variety of wells and configurations. Thermo Scientific™ Immuno Breakable Modules offer an optimal choice of products for the specific assay. They offer lot-to-lot reliability and well-to-well reproducibility.
Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.- For solid phase immuno assays, e.g. ELISA
- Choice of four surfaces for optimal binding
- Free-standing modules are easy to handle
- Alphanumeric system for easy orientation
- Fit standard equipment
- High optical quality for reliable results
- Polystyrene
Choice of Surface
- PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
- Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
- MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
- MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Specifications
Breakable Strips | |
Yes | |
Without Lid | |
Colorimetric | |
60 Cases | |
Frame | |
350 μL | |
250 μL | |
Round, Curved Edges, Flat Bottom (C-well) | |
60 | |
None |
Slightly Hydrophilic | |
Strip Plate | |
High optical quality for reliable results | |
8 x 12 | |
Clear | |
Polystyrene | |
96 | |
MediSorp | |
For binding a diverse range of biomolecules | |
10 | |
Proteins |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185229 | Certificate of Compliance | 15/02/2025 | 446470 |
185061 | Certificate of Compliance | 23/01/2025 | 446470 |
163573 | Certificate of Compliance | 03/12/2024 | 446470 |
166337 | Certificate of Compliance | 03/12/2024 | 446470 |
172804 | Certificate of Compliance | 03/12/2024 | 446470 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.