Learn More
Fisherbrand™ Insert for Midi Boxes
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
Brand:  Fisherbrand™ CSR-RDUO
 View more versions of this product
Product Code. 12387167
  • 46.80 € / Each
Estimated Shipment: 21-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Microcentrifuge tube beta for FB58447/FB58448
  • 16 x 1.5mL and 16 x 0.5mL tubes 160 x 80 x 40mm
  • Various inserts to fit inside Midi boxes
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
For Maxi Boxes
Insert
160mm
40mm
16 x 1.5/16 x 0.5mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Insert for Midi Boxes > Dimensions: 16 x 1.5 and 16 x 0.5ml tubes