missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ gDNA Plant Kit
GREENER_CHOICE

The ChargeSwitch™ gDNA Plant Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from fungal and plant samples, including leaves, tissues and seeds, in less than 15 minutes after lysate preparation.

Brand:  Invitrogen™ CS18000

 View more versions of this product

Product Code. 10695273

  • 474.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch Lysis Buffer (L18)
ChargeSwitch Precipitation Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E6; 10 mM Tris-HCl, pH 8.5 with 1 mM EDTA)
RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
10% SDS in deionized water
10% Detergent (D1)
This item is not returnable. View return policy

Description

Description

The ChargeSwitch™ gDNA Plant Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from fungal and plant samples, including leaves, tissues and seeds, in less than 15 minutes after lysate preparation.

• Optimized for higher-purity DNA extraction
• Compatible with high-throughput protocols
• Ideal for a variety of downstream applications
• Less hazardous, less waste

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2945738 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberCS18000
Lot Number3088736 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberCS18000
Lot Number2399260 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCS18000
Lot Number2463687 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCS18000
Lot Number2579979 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCS18000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ChargeSwitch™ gDNA Plant Kit > 96 preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.