Learn More
Invitrogen™ Total Exosome Isolation Reagent (from urine)
Total Exosome Isolation Reagent from urine enables fast and efficient enrichment of intact exosomes with a protocol that is scalable depending on sample size.
Brand: Invitrogen™ 4484452
276.25 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Total Exosome Isolation Reagent from urine enables fast and efficient enrichment of intact exosomes with a protocol that is scalable depending on sample size.
- Maximize recovery of intact exosomes for any type of downstream application
- Avoid time-consuming ultra-centrifugation
- Maximize flexibility—can be scaled up or down depending on the sample size

Specifications
Store at 2–8°C. | |
Exosomes | |
50 mL |
Total Exosome Isolation | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2802872 | Certificate of Analysis | 29/01/2025 | 4484452 |
2855527 | Certificate of Analysis | 28/01/2025 | 4484452 |
3094372 | Certificate of Analysis | 27/01/2025 | 4484452 |
2879296 | Certificate of Analysis | 26/01/2025 | 4484452 |
2964557 | Certificate of Analysis | 26/01/2025 | 4484452 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.