missing translation for 'onlineSavingsMsg'
Learn More
Isobutyl benzoate, 99%, Thermo Scientific™ Product Code.: 11458870

Isobutyl benzoate, 99%, Thermo Scientific™

Product Code. 11458870
25 g
Click to view available options
Quantity:
25 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11458870

Brand: Thermo Scientific Alfa Aesar A10114.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-50-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00048344
InChI Key KYZHGEFMXZOSJN-UHFFFAOYSA-N
Synonym isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoateShow MoreShow Less
PubChem CID 61048
ChEBI CHEBI:87500
IUPAC Name 2-methylpropyl benzoate
SMILES CC(C)COC(=O)C1=CC=CC=C1

Specifications

Density 0.994
Boiling Point 242°C
Flash Point 96°C (204°F)
Refractive Index 1.494
Quantity 25 g
Beilstein 2045961
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material Isobutyl benzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10127515 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10174804 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isobutyl benzoate, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.