missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A40 Light Duty Overboots
Click to view available options
Size: Universal
Description
- Single-use light duty overboots
- Ideal for keeping footwear clean and covered
- Reduce contamination in food processing and light industrial areas
- PPE Category I - CE Simple
Specifications
Specifications
Size | Universal |
Material | Polyethylene |
Latex Free | Yes |
Coating | None |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic with Ties |
Sole Type | Anti-slip |
For Use With (Application) | Food Processing, Light Industrial |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kimberly-Clark Professional™ KleenGuard™ A40 Light Duty Overboots >
Spot an opportunity for improvement?Share a Content Correction