missing translation for 'onlineSavingsMsg'
Learn More
L-Isoleucine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 15401929

L-Isoleucine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15401929
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15401929

Brand: Thermo Scientific Alfa Aesar H64640.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56782-52-6
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.69
MDL Number MFCD09953729
InChI Key QQGRWNMNWONMOO-LEUCUCNGSA-N
Synonym l-isoleucine ethyl ester hydrochloride, h-ile-oet.hcl, ethyl l-isoleucinate hydrochloride, l-isoleucine ethyl ester hcl, 2s,3s-ethyl 2-amino-3-methylpentanoate hydrochloride, isoleucine, ethyl ester, hydrochloride, ethyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, l-isoleucine ethylester hcl, ksc497o6p, l-isoleucineethylesterhydrochlorideShow MoreShow Less
PubChem CID 6453390
SMILES Cl.CCOC(=O)[C@@H](N)[C@@H](C)CC

Specifications

Color White
Beilstein 4710859
IUPAC Name hydrogen ethyl (2S,3S)-2-amino-3-methylpentanoate chloride
Quantity 5 g
Formula Weight 195.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Isoleucine ethyl ester hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM27D045 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH64640.06,H64640.14
Lot Number81400045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64640.06,H64640.14
Lot NumberQ22H052 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64640.06,H64640.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Isoleucine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.