missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Lab Jack with Metal Knob

Bochem™ Lab Jack with Metal Knob that is ergonomic, rounded, and user-friendly.

Brand:  Bochem™ 11133

 View more versions of this product

Product Code. 15881797

  • 264.00 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • All models are manufactured according to DIN 12897
  • Improved scissor technology with reduced tolerances
  • Ergonomic knob, rounded and user-friendly
  • Scissor-type design made of 18/10 steel
  • Maximum dynamic operating load: 7kg
  • Maximum static operating load 30kg
Specifications

Specifications

Stainless Steel (18/10)
200 mm
M8 x 1.25
60 to 275 mm
200 mm
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ Lab Jack with Metal Knob > Length: 200mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.