missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester GHS Symbol Signs

Help ensure safety at work and maximize productivity with these GHS warning labels. Brady™ Laminated Polyester GHS Symbol Signs are available in a variety of sizes, with and without text. Standardized images reduce the risk of misunderstandings.

Brand:  Brady™ 811694

 View more versions of this product

Product Code. 11548884

This item is not returnable. View return policy

Description

Description

  • Self-adhesive diamond shape labels
  • Pre-printed with GHS hazard symbols
  • Laminated polyester, gloss finish black and red on white
Specifications

Specifications

ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Gloss
250/Pk.
Permanent Acrylic, Pressure-sensitive
Pipe markers and safety signs
0.024 mm
-40°C to 110°C
Compressed Gas
Black on White
PIC 1804-100*100-B7541-RLL
Polyester
Individual Markers on a Roll
Laminated Polyester
100 mm
100 mm
Diamond
GHS Symbol Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Laminated Polyester GHS Symbol Signs > Compressed Gas; Width: 100mm; 250 Pack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.