missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mini Gel Tank

Compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels

Brand:  Invitrogen™ A25977

Product Code. 15320424

  • 491.00 € / Each
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket

Includes: Gel Runner Tank, Mini Gel Tank Base, Mini Gel Tank Lid, Cassette Clamps, right and left, Novex™ Power Supply Adapters, Gel Knife
This item is not returnable. View return policy

Description

Description

Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.

The Mini Gel Tank offers: 
 

  • Versatility—compatible with NuPAGE™, Novex™, and Bolt mini gels
  • Easy sample loading—forward-facing well configuration
  • Simultaneous visualization of both gels—streamlined, side-by-side tank configuration
  • Simple monitoring of prestained protein markers— with white tank stand
  • Less running buffer required—two separate gel chambers, so you only need to load sufficient buffer for each gel to the specified fill line
  • High performance—same great results you've been getting with the XCell SureLock™ Mini-Cell and original Bolt Mini Gel Tank
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Includes Runner Tank, Cassette Clamps (left & right), Tank Base, Tank Lid, and Power Supply Adapters.
Bolt™ Bis-Tris Plus Gels, Novex™ Mini Gels
35 min.
1 unit
500 VDC
2 Gels/Run
Mini
Mini Gel Tank
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number110840001 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberA25977,A25977_LITL18 PROMO
Lot Number110940002 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberA25977,A25977_LITL18 PROMO
Lot Number110260001 Certificate TypeCertificate of Analysis Date17/04/2017 Catalog NumberA25977,A25977_LITL18 PROMO
Lot Number110450008 Certificate TypeCertificate of Analysis Date16/04/2017 Catalog NumberA25977,A25977_LITL18 PROMO
Lot Number110250004 Certificate TypeCertificate of Analysis Date16/04/2017 Catalog NumberA25977,A25977_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Mini Gel Tank > 1 unit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.