missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Low-Density Polyethylene Bags

Store a variety of solids or powders in these LDPE bags. Low Density Polyethylene Bags stand up to your storage needs with a strong bottom seal.

Brand:  Cole-Parmer™ 06505-14

 View more versions of this product

Product Code. 12303820

  • 131.00 € / Pack of 500
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Seal with heat sealer or bag ties
  • Good for general laboratory storage
  • Available in a variety of sizes and thicknesses
Specifications

Specifications

Low-Density Bag
Polyethylene
0.03 mil
254 mm
Clear
500 Each
254 x 203mm
203 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Low-Density Polyethylene Bags > Dimensions (L x W): 254 x 203 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.